Gene Transcripts

Gene Symbol
NLGN4Y
Entrez Gene ID
22829
Gene Name
neuroligin 4, Y-linked
Gene Aliases
Chromosome Location
Chr. Y: 14522578 - 14844945 on Build GRCh38
UniGene ID
Hs.439199
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164238.1 NP_001157710.1 1 456
NM_014893.4 NP_055708.3 2 609
NR_028319.1 2 831
NR_046355.1 2 536
XM_006724874.2 XP_006724937.1 2 781
XM_011531424.2 XP_011529726.1 2 1099
XM_011531425.2 XP_011529727.1 2 871
XM_011531426.2 XP_011529728.1 2 832
XM_011531427.1 XP_011529729.1 2 586
XM_011531428.1 XP_011529730.1 3 767
XM_011531429.2 XP_011529731.1 2 1382
XM_011531430.2 XP_011529732.1 2 720
XM_017030034.1 XP_016885523.1 2 871
XM_017030035.1 XP_016885524.1 2 792
XM_017030036.1 XP_016885525.1 2 586
XM_017030037.1 XP_016885526.1 2 584
XM_017030038.1 XP_016885527.1 2 1382
XM_017030041.1 XP_016885530.1 2 870
GenBank mRNA AF376804.1 - 2 609
AK295117.1 - 2 536
BC032567.1 - 1 259
BC113525.1 - 1 436 8322577
BC113551.1 - 1 436 8322603
BX537428.1 - 2 832

View Full Product Details