Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001281302.1 | NP_001268231.1 | 16 | 1993 | |
NM_032380.4 | NP_115756.2 | 15 | 1875 | ||
NM_170681.2 | NP_733781.1 | 15 | 1875 | ||
NM_170691.2 | NP_733792.1 | 14 | 1734 | ||
NR_104006.1 | 15 | 1875 | |||
XM_011543691.2 | XP_011541993.1 | 15 | 1658 | ||
XM_017009986.1 | XP_016865475.1 | 15 | 1769 | ||
GenBank mRNA | AF111808.1 | - | 14 | 1484 | |
AF367997.1 | - | 15 | 1721 | ||
AK025314.1 | - | 15 | 1652 | ||
AK315705.1 | - | 15 | 1654 | ||
AY453587.1 | - | 15 | 1619 | ||
BC015712.1 | - | 16 | 1727 | 3922036 | |
BC030612.1 | - | 15 | 1624 | 4811939 |