Gene Transcripts
- Gene Symbol
- STK19
- Entrez Gene ID
- 8859
- Gene Name
- serine/threonine kinase 19
- Gene Aliases
- D6S60, D6S60E, G11, HLA-RP1, RP1
- Chromosome Location
- Chr. 6: 31971175 - 31981446 on Build GRCh38
- UniGene ID
- Hs.654371
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_004197.1 |
NP_004188.1
|
5, 6 |
924 |
|
NM_032454.1 |
NP_115830.1
|
5, 6 |
936 |
|
NR_026717.1 |
|
5, 6 |
1109 |
|
GenBank mRNA |
BC110347.1 |
- |
5, 6 |
1109 |
|
BC121128.1 |
- |
4, 5 |
943 |
40120738 |
BC121129.1 |
- |
4, 5 |
942 |
40120740 |
BC143833.1 |
- |
4, 5 |
979 |
9052348 |
BC143835.1 |
- |
4, 5 |
991 |
|
L26260.1 |
- |
5, 6 |
924 |
|
X77386.1 |
- |
4, 5 |
530 |
|
View Full Product Details