Gene Transcripts

Gene Symbol
SBNO2
Entrez Gene ID
22904
Gene Name
strawberry notch homolog 2
Gene Aliases
KIAA0963, SNO, STNO
Chromosome Location
Chr. 19: 1107634 - 1174283 on Build GRCh38
UniGene ID
Hs.408708
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100122.1 NP_001093592.1 13, 14 1718
NM_014963.2 NP_055778.2 16, 17 2050
XM_011527804.2 XP_011526106.1 16, 17 2023
GenBank mRNA AB023180.1 - 16, 17 2027
AK074102.1 - 12, 13 1510
AK125139.1 - 13, 14 1665
AK292407.1 - 16, 17 2050
AK296849.1 - 13, 14 1665
AK300981.1 - 17, 18 1990
AK309167.1 - 13, 14 1665
BG332816.1 - 2, 3 235

View Full Product Details