Gene Transcripts

Gene Symbol
CA12
Entrez Gene ID
771
Gene Name
carbonic anhydrase 12
Gene Aliases
CA-XII, CAXII, HsT18816, T18816
Chromosome Location
Chr. 15: 63321378 - 63382166 on Build GRCh38
UniGene ID
Hs.210995
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001218.4 NP_001209.1 5 847
NM_001293642.1 NP_001280571.1 4 667
NM_206925.2 NP_996808.1 5 847
NR_135511.1 5 950
GenBank mRNA AF037335.1 - 5 572
AF051882.1 - 5 464
AK096845.1 - 4 403
AK315769.1 - 5 613
BC000278.1 - 5 569 3357439
BC011691.2 - 5 572 3344734
BC023981.1 - 5 587 4042589
BT006656.1 - 5 457

View Full Product Details