Gene Transcripts

Gene Symbol
OVOS
Entrez Gene ID
408186
Gene Name
ovostatin
Gene Aliases
Chromosome Location
Chr. 12: 9517803 - 9573287 on Build GRCh38
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017020287.1 XP_016875776.1 9, 10 1009

View Full Product Details