Gene Transcripts
- Gene Symbol
- YIF1A
- Entrez Gene ID
- 10897
- Gene Name
- Yip1 interacting factor homolog A, membrane trafficking protein
- Gene Aliases
- 54TM, FinGER7, YIF1, YIF1P
- Chromosome Location
- Chr. 11: 66284580 - 66289167 on Build GRCh38
- UniGene ID
- Hs.446445
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001300861.1 |
NP_001287790.1
|
4, 5 |
618 |
|
NM_020470.2 |
NP_065203.2
|
4, 5 |
618 |
|
XM_005273720.3 |
XP_005273777.1
|
4, 5 |
594 |
|
XM_017017139.1 |
XP_016872628.1
|
4, 5 |
594 |
|
XM_017017140.1 |
XP_016872629.1
|
4, 5 |
594 |
|
GenBank mRNA |
AK226003.1 |
- |
4, 5 |
596 |
|
AK291124.1 |
- |
4, 5 |
596 |
|
BC001299.1 |
- |
4, 5 |
569 |
3451489 |
BC009892.2 |
- |
1, 2 |
45 |
|
BG397210.1 |
- |
4, 5 |
525 |
|
CR456710.1 |
- |
4, 5 |
436 |
|
View Full Product Details