Gene Transcripts
- Gene Symbol
- TBC1D3L
- Entrez Gene ID
- 101060376
- Gene Name
- TBC1 domain family member 3L
- Gene Aliases
- Chromosome Location
- Chr. 17: 37978155 - 37990691 on Build GRCh38
- UniGene ID
- Hs.454716
- Species
- Homo sapiens
- Gene Symbol
- LOC101060389
- Entrez Gene ID
- 101060389
- Gene Name
- TBC1 domain family member-like
- Gene Aliases
- TBC1D3D
- Chromosome Location
- Chr. 17: 38004033 - 38014908 on Build GRCh38
- UniGene ID
- Hs.454716
- Species
- Homo sapiens
- Gene Symbol
- TBC1D3
- Entrez Gene ID
- 729873
- Gene Name
- TBC1 domain family member 3
- Gene Aliases
- PRC17, TBC1D3A, TBC1D3F
- Chromosome Location
- Chr. 17: 38181476 - 38192555 on Build GRCh38
- UniGene ID
- Hs.454716
- Species
- Homo sapiens
- Gene Symbol
- TBC1D3B
- Entrez Gene ID
- 414059
- Gene Name
- TBC1 domain family member 3B
- Gene Aliases
- PRC17, TBC1D3I, TBC1D3L
- Chromosome Location
- Chr. 17: 36165681 - 36176624 on Build GRCh38
- UniGene ID
- Hs.105891
- Species
- Homo sapiens
- Gene Symbol
- TBC1D3C
- Entrez Gene ID
- 414060
- Gene Name
- TBC1 domain family member 3C
- Gene Aliases
- TBC1D3D
- Chromosome Location
- Chr. 17: 38057693 - 38068634 on Build GRCh38
- UniGene ID
- Hs.735975
- Species
- Homo sapiens
- Gene Symbol
- TBC1D3F
- Entrez Gene ID
- 84218
- Gene Name
- TBC1 domain family member 3F
- Gene Aliases
- TBC1D3
- Chromosome Location
- Chr. 17: 36428612 - 36439523 on Build GRCh38
- UniGene ID
- Hs.617010
- Species
- Homo sapiens
- Gene Symbol
- TBC1D3G
- Entrez Gene ID
- 101060321
- Gene Name
- TBC1 domain family member 3G
- Gene Aliases
- Chromosome Location
- Chr. 17: 36322242 - 36334759 on Build GRCh38
- UniGene ID
- Hs.454716
- Species
- Homo sapiens
- Gene Symbol
- TBC1D3H
- Entrez Gene ID
- 729877
- Gene Name
- TBC1 domain family member 3H
- Gene Aliases
- Chromosome Location
- Chr. 17: 36377475 - 36388448 on Build GRCh38
- UniGene ID
- Hs.105891
- Species
- Homo sapiens
- Gene Symbol
- TBC1D3I
- Entrez Gene ID
- 102724862
- Gene Name
- TBC1 domain family member 3I
- Gene Aliases
- Chromosome Location
- Chr. 17: 36253639 - 36264601 on Build GRCh38
- UniGene ID
- Hs.454716
- Species
- Homo sapiens
- Gene Symbol
- TBC1D3J
- Entrez Gene ID
- 100510707
- Gene Name
- TBC1 domain family member 3J
- Gene Aliases
- Chromosome Location
- Chr. 17: 36450649 - 36458000 on Build GRCh38
- Species
- Homo sapiens
- Gene Symbol
- TBC1D3K
- Entrez Gene ID
- 101060351
- Gene Name
- TBC1 domain family member 3K
- Gene Aliases
- Chromosome Location
- Chr. 17: 37923805 - 37935371 on Build GRCh38
- UniGene ID
- Hs.454716
- Species
- Homo sapiens
View Full Product Details