Gene Transcripts

Gene Symbol
Lrrd1
Entrez Gene ID
242838
Gene Name
leucine rich repeats and death domain containing 1
Gene Aliases
4932412H11Rik
Chromosome Location
Chr. 5: 3845173 - 3866596 on Build GRCm38
UniGene ID
Mm.482273
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172879.3 NP_766467.2 2 1084
GenBank mRNA AK029981.1 - 2 1085
BC141026.1 - 2 911 9055638
BC145170.1 - 2 911 9053721

View Full Product Details