Gene Transcripts

Gene Symbol
Osbpl7
Entrez Gene ID
71240
Gene Name
oxysterol binding protein-like 7
Gene Aliases
4933437E18Rik
Chromosome Location
Chr. 11: 97050344 - 97068904 on Build GRCm38
UniGene ID
Mm.190450
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081434.1 NP_001074903.1 14 1492
GenBank mRNA AK154677.1 - 14 1528
AK171585.1 - 14 1521

View Full Product Details