Gene Transcripts

Gene Symbol
Trappc10
Entrez Gene ID
216131
Gene Name
trafficking protein particle complex 10
Gene Aliases
B230307C21Rik, Gm870, Tmem1
Chromosome Location
Chr. 10: 78186725 - 78244642 on Build GRCm38
UniGene ID
Mm.27539
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081055.1 NP_001074524.1 21, 22 3475
GenBank mRNA AK154290.1 - 21, 22 3412
AK185223.1 - 1, 2 77
BC044902.1 - 13, 14 2131

View Full Product Details