Gene Transcripts

Gene Symbol
Prr23a
Entrez Gene ID
623166
Gene Name
proline rich 23A
Gene Aliases
EG623166, Gm6400
Chromosome Location
Chr. 9: 98842587 - 98843645 on Build GRCm38
UniGene ID
Mm.390211
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134660.1 NP_001128132.1 1 979

View Full Product Details