Gene Transcripts

Gene Symbol
Vmn2r51
Entrez Gene ID
100042921
Gene Name
vomeronasal 2, receptor 51
Gene Aliases
ENSMUSG00000058685
Chromosome Location
Chr. 7: 10087198 - 10105659 on Build GRCm38
UniGene ID
Mm.291126
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105179.1 NP_001098649.1 2 339

View Full Product Details