Gene Transcripts

Gene Symbol
Nr1i2
Entrez Gene ID
18171
Gene Name
nuclear receptor subfamily 1, group I, member 2
Gene Aliases
PXR, PXR.1, PXR.2, PXR1, SXR, mPXR
Chromosome Location
Chr. 16: 38248321 - 38294851 on Build GRCm38
UniGene ID
Mm.8509
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001098404.1 NP_001091874.1 2 417
NM_010936.3 NP_035066.1 2 417
GenBank mRNA AF031814.1 - 2 232
AI746915.1 - 2 417
AK011108.1 - 2 386
AK018630.1 - 2 417
BC120796.1 - 2 164 40129719
BC137780.1 - 2 164 8860799

View Full Product Details