Gene Transcripts
- Gene Symbol
- Hist1h2bp
- Entrez Gene ID
- 319188
- Gene Name
- histone cluster 1, H2bp
- Gene Aliases
- Chromosome Location
- Chr. 13: 21787480 - 21801561 on Build GRCm38
- UniGene ID
- Mm.264645
- Species
- Mus musculus
- Gene Symbol
- Hist1h2bb
- Entrez Gene ID
- 319178
- Gene Name
- histone cluster 1, H2bb
- Gene Aliases
- H2b-143
- Chromosome Location
- Chr. 13: 23746734 - 23747223 on Build GRCm38
- UniGene ID
- Mm.390567
- Species
- Mus musculus
- Gene Symbol
- Hist1h2bc
- Entrez Gene ID
- 68024
- Gene Name
- histone cluster 1, H2bc
- Gene Aliases
- 2610022J01Rik, H2bfs, R74621
- Chromosome Location
- Chr. 13: 23684199 - 23692480 on Build GRCm38
- UniGene ID
- Mm.261676
- Species
- Mus musculus
- Gene Symbol
- Hist1h2be
- Entrez Gene ID
- 319179
- Gene Name
- histone cluster 1, H2be
- Gene Aliases
- Chromosome Location
- Chr. 13: 23583670 - 23621124 on Build GRCm38
- UniGene ID
- Mm.440391
- Species
- Mus musculus
- Gene Symbol
- Hist1h2bg
- Entrez Gene ID
- 319181
- Gene Name
- histone cluster 1, H2bg
- Gene Aliases
- Chromosome Location
- Chr. 13: 23571400 - 23571863 on Build GRCm38
- UniGene ID
- Mm.261673
- Species
- Mus musculus
- Gene Symbol
- Hist1h2bh
- Entrez Gene ID
- 319182
- Gene Name
- histone cluster 1, H2bh
- Gene Aliases
- H2b-221
- Chromosome Location
- Chr. 13: 23542923 - 23543444 on Build GRCm38
- UniGene ID
- Mm.352273
- Species
- Mus musculus
- Gene Symbol
- Hist1h2bj
- Entrez Gene ID
- 319183
- Gene Name
- histone cluster 1, H2bj
- Gene Aliases
- Chromosome Location
- Chr. 13: 22043230 - 22043658 on Build GRCm38
- UniGene ID
- Mm.371766
- Species
- Mus musculus
- Gene Symbol
- Hist1h2bk
- Entrez Gene ID
- 319184
- Gene Name
- histone cluster 1, H2bk
- Gene Aliases
- Chromosome Location
- Chr. 13: 22035821 - 22036320 on Build GRCm38
- UniGene ID
- Mm.358955
- Species
- Mus musculus
- Gene Symbol
- Hist1h2bl
- Entrez Gene ID
- 319185
- Gene Name
- histone cluster 1, H2bl
- Gene Aliases
- H2b-291a
- Chromosome Location
- Chr. 13: 21715713 - 21716143 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_178199.2 |
NP_835506.1
|
1 |
193 |
|
- Gene Symbol
- Hist1h2bm
- Entrez Gene ID
- 319186
- Gene Name
- histone cluster 1, H2bm
- Gene Aliases
- H2b-291b
- Chromosome Location
- Chr. 13: 21722044 - 21722526 on Build GRCm38
- UniGene ID
- Mm.277425
- Species
- Mus musculus
- Gene Symbol
- Hist1h2bn
- Entrez Gene ID
- 319187
- Gene Name
- histone cluster 1, H2bn
- Gene Aliases
- Chromosome Location
- Chr. 13: 21754123 - 21754553 on Build GRCm38
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_178201.2 |
NP_835508.1
|
1 |
193 |
|
- Gene Symbol
- Hist1h2bq
- Entrez Gene ID
- 665596
- Gene Name
- histone cluster 1, H2bq
- Gene Aliases
- Gm13646, OTTMUSG00000013203
- Chromosome Location
- Chr. 13: 21806412 - 21810199 on Build GRCm38
- UniGene ID
- Mm.371766
- Species
- Mus musculus
- Gene Symbol
- Hist1h2br
- Entrez Gene ID
- 665622
- Gene Name
- histone cluster 1 H2br
- Gene Aliases
- Gm11277, OTTMUSG00000000449
- Chromosome Location
- Chr. 13: 21833743 - 21837530 on Build GRCm38
- UniGene ID
- Mm.471247
- Species
- Mus musculus
View Full Product Details