Gene Transcripts

Gene Symbol
Olfr27
Entrez Gene ID
258826
Gene Name
olfactory receptor 27
Gene Aliases
MOR171-6, MTPCR56
Chromosome Location
Chr. 9: 39128167 - 39145072 on Build GRCm38
UniGene ID
Mm.246532
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_146829.2 NP_667040.2 2 576

Gene Symbol
Olfr1537
Entrez Gene ID
257959
Gene Name
olfactory receptor 1537
Gene Aliases
K4, MOR171-32P, MOR171-41P, Olfr144, Olfr1537-ps1
Chromosome Location
Chr. 9: 39237487 - 39238431 on Build GRCm38
UniGene ID
Mm.335076
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207665.1 NP_997548.1 1 515

View Full Product Details