Gene Transcripts

Gene Symbol
PACS2
Entrez Gene ID
23241
Gene Name
phosphofurin acidic cluster sorting protein 2
Gene Aliases
PACS-2, PACS1L
Chromosome Location
Chr. 14: 105300811 - 105398147 on Build GRCh38
UniGene ID
Hs.525626
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100913.2 NP_001094383.2 16 2187
NM_001243127.2 NP_001230056.1 16 1697
NM_015197.3 NP_056012.2 16 2175
XM_006720090.1 XP_006720153.1 16 1855
XM_006720091.1 XP_006720154.1 16 1855
XM_006720092.2 XP_006720155.1 16 1816
XM_017021105.1 XP_016876594.1 16 1843
XM_017021106.1 XP_016876595.1 16 1840
XM_017021107.1 XP_016876596.1 16 1831
XM_017021108.1 XP_016876597.1 16 1828
XM_017021109.1 XP_016876598.1 16 1852
XM_017021110.1 XP_016876599.1 16 1843
XM_017021111.1 XP_016876600.1 16 1721
XM_017021112.1 XP_016876601.1 16 1623
GenBank mRNA AB011174.1 - 16 1858
AY320284.2 - 16 2174
BC010663.1 - 13 1282
BC050351.1 - 16 1713 5264871
BC065220.1 - 16 1788 5732786
BC131591.1 - 16 1675

View Full Product Details