Gene Transcripts

Gene Symbol
Vps13c
Entrez Gene ID
320528
Gene Name
vacuolar protein sorting 13C (yeast)
Gene Aliases
AI552542, AV026040, C230055H22Rik, W51662, chorein
Chromosome Location
Chr. 9: 67840331 - 67995640 on Build GRCm38
UniGene ID
Mm.241454
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177184.3 NP_796158.2 24 2363
GenBank mRNA AB281675.1 - 24 2363
AK083667.1 - 1 2118

View Full Product Details