Gene Transcripts

Gene Symbol
Rsph4a
Entrez Gene ID
309767
Gene Name
radial spoke head 4 homolog A (Chlamydomonas)
Gene Aliases
Rshl3
Chromosome Location
Chr. 20: 32450701 - 32467339 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003751961.3 XP_003752009.1 1, 2 831

Gene Symbol
LOC103689994
Entrez Gene ID
103689994
Gene Name
radial spoke head protein 4 homolog A
Gene Aliases
Chromosome Location
Chr. 20: 33945770 - 33962054 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003751965.3 XP_003752013.1 831

View Full Product Details