Gene Transcripts

Gene Symbol
Maff
Entrez Gene ID
366960
Gene Name
v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog F
Gene Aliases
Chromosome Location
Chr. 7: 120580743 - 120592095 on Build Rnor_6.0
UniGene ID
Rn.31639
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130573.1 NP_001124045.1 3 687
XM_006242029.2 XP_006242091.2 3 1092

View Full Product Details