Gene Transcripts

Gene Symbol
RIMBP2
Entrez Gene ID
23504
Gene Name
RIMS binding protein 2
Gene Aliases
PPP1R133, RBP2, RIM-BP2
Chromosome Location
Chr. 12: 130396133 - 130716361 on Build GRCh38
UniGene ID
Hs.657441
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015347.4 NP_056162.4 15 2899
XM_011538102.1 XP_011536404.1 17 3717
XM_011538103.2 XP_011536405.1 19 4179
XM_011538104.2 XP_011536406.1 19 3994
XM_011538105.2 XP_011536407.1 18 3451
XM_011538106.2 XP_011536408.1 17 3292
XM_017019105.1 XP_016874594.1 19 4192
XM_017019106.1 XP_016874595.1 18 3451
XM_017019107.1 XP_016874596.1 18 3273
XM_017019108.1 XP_016874597.1 18 3456
XM_017019109.1 XP_016874598.1 16 3000
XM_017019110.1 XP_016874599.1 18 3273
XM_017019111.1 XP_016874600.1 18 3273
GenBank mRNA AB002316.1 - 15 2899
AY699979.1 - 12 2590
BX641152.1 - 2 7631

View Full Product Details