Gene Transcripts

Gene Symbol
MXRA5
Entrez Gene ID
25878
Gene Name
matrix remodeling associated 5
Gene Aliases
Chromosome Location
Chr. X: 3308565 - 3347098 on Build GRCh38
UniGene ID
Hs.369422
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015419.3 NP_056234.2 5 1298
XM_005274485.2 XP_005274542.1 5 1753
GenBank mRNA AF245505.1 - 4 1141
BC011846.2 - 5 1295
BC064986.1 - 5 1295
BC080586.1 - 5 1295

View Full Product Details