Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001243259.1 | NP_001230188.1 | 10 | 1469 | |
NM_015517.4 | NP_056332.2 | 11 | 1524 | ||
NM_198971.2 | NP_945322.1 | 10 | 1407 | ||
XM_011542744.2 | XP_011541046.1 | 10 | 1516 | ||
XM_011542745.2 | XP_011541047.1 | 10 | 1366 | ||
XM_011542746.1 | XP_011541048.1 | 9 | 1187 | ||
XM_017017499.1 | XP_016872988.1 | 10 | 1604 | ||
XM_017017500.1 | XP_016872989.1 | 10 | 1483 | ||
XM_017017501.1 | XP_016872990.1 | 11 | 1610 | ||
XM_017017502.1 | XP_016872991.1 | 13 | 1795 | ||
XM_017017503.1 | XP_016872992.1 | 11 | 1560 | ||
XM_017017504.1 | XP_016872993.1 | 11 | 1435 | ||
XM_017017505.1 | XP_016872994.1 | 11 | 1413 | ||
XM_017017506.1 | XP_016872995.1 | 11 | 1376 | ||
XM_017017507.1 | XP_016872996.1 | 13 | 1615 | ||
XM_017017508.1 | XP_016872997.1 | 12 | 1490 | ||
XM_017017509.1 | XP_016872998.1 | 10 | 1242 | ||
GenBank mRNA | AK056362.1 | - | 10 | 1320 | |
AK092869.1 | - | 5 | 1661 | ||
AK301452.1 | - | 10 | 1376 | ||
AL080201.1 | - | 2 | 217 | ||
BC001073.2 | - | 10 | 1378 | 2820431 | |
BC001945.1 | - | 10 | 1378 | 2820431 | |
BC012856.1 | - | 10 | 1319 | 3871231 | |
BC017234.2 | - | 10 | 1327 | 3537381 |