Gene Transcripts

Gene Symbol
RGS22
Entrez Gene ID
26166
Gene Name
regulator of G-protein signaling 22
Gene Aliases
CT145, PRTD-NY2
Chromosome Location
Chr. 8: 99960936 - 100106116 on Build GRCh38
UniGene ID
Hs.120021
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286692.1 NP_001273621.1 19 3035
NM_001286693.1 NP_001273622.1 17 2930
NM_015668.4 NP_056483.3 19 3071
XM_005250857.2 XP_005250914.1 19 2917
XM_005250858.2 XP_005250915.1 19 2917
XM_006716540.2 XP_006716603.1 19 3185
XM_011516957.1 XP_011515259.1 20 3366
XM_011516958.1 XP_011515260.1 19 3264
XM_017013311.1 XP_016868800.1 12 1614
GenBank mRNA AK292559.1 - 19 2881
AY009106.1 - 19 2864
BC036665.1 - 19 2922 5268331
BC092411.1 - 19 3068 30386231

View Full Product Details