Gene Transcripts
- Gene Symbol
- VENTXP7
- Entrez Gene ID
- 391518
- Gene Name
- VENT homeobox pseudogene 7
- Gene Aliases
- HPX42, VENTX1
- Chromosome Location
- Chr. 3: 21405726 - 21406708 on Build GRCh38
- UniGene ID
- Hs.543226
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002311.1 |
|
1 |
393 |
|
GenBank mRNA |
X74864.1 |
- |
1 |
91 |
|
- Gene Symbol
- VENTX
- Entrez Gene ID
- 27287
- Gene Name
- VENT homeobox
- Gene Aliases
- HPX42B, NA88A, VENTX2
- Chromosome Location
- Chr. 10: 133237904 - 133241930 on Build GRCh38
- UniGene ID
- Hs.125231
- Species
- Homo sapiens
View Full Product Details