Gene Transcripts

Gene Symbol
SCHIP1
Entrez Gene ID
29970
Gene Name
schwannomin interacting protein 1
Gene Aliases
SCHIP-1
Chromosome Location
Chr. 3: 159273247 - 159897366 on Build GRCh38
UniGene ID
Hs.134665
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001197107.1 NP_001184036.1 1, 2 636
NM_014575.3 NP_055390.1 1, 2 636
GenBank mRNA AF145713.1 - 1, 2 131
AF145715.1 - 1, 2 131
AK314662.1 - 1, 2 579
BC036535.2 - 1, 2 639 5263291

View Full Product Details