Gene Transcripts
- Gene Symbol
- SCHIP1
- Entrez Gene ID
- 29970
- Gene Name
- schwannomin interacting protein 1
- Gene Aliases
- SCHIP-1
- Chromosome Location
- Chr. 3: 159273247 - 159897366 on Build GRCh38
- UniGene ID
- Hs.134665
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001197107.1 |
NP_001184036.1
|
4, 5 |
1542 |
|
NM_001197108.1 |
NP_001184037.1
|
3, 4 |
885 |
|
NM_001197109.1 |
NP_001184038.1
|
3, 4 |
602 |
|
NM_014575.3 |
NP_055390.1
|
4, 5 |
1581 |
|
GenBank mRNA |
AF070614.1 |
- |
3, 4 |
691 |
|
AF145713.1 |
- |
4, 5 |
1076 |
|
AF145714.1 |
- |
3, 4 |
380 |
|
AF145715.1 |
- |
4, 5 |
1037 |
|
AK091871.1 |
- |
3, 4 |
523 |
|
AK309223.1 |
- |
4, 5 |
989 |
|
AK314662.1 |
- |
4, 5 |
1524 |
|
BC005947.1 |
- |
3, 4 |
418 |
4246326 |
BC036535.2 |
- |
4, 5 |
1581 |
5263291 |
BX648179.1 |
- |
3 |
2099 |
|
- Gene Symbol
- IQCJ-SCHIP1
- Entrez Gene ID
- 100505385
- Gene Name
- IQCJ-SCHIP1 readthrough
- Gene Aliases
- Chromosome Location
- Chr. 3: 159069252 - 159897366 on Build GRCh38
- UniGene ID
- Hs.134665
- Species
- Homo sapiens
View Full Product Details