Gene Transcripts

Gene Symbol
TAS2R7
Entrez Gene ID
50837
Gene Name
taste 2 receptor member 7
Gene Aliases
T2R7, TRB4
Chromosome Location
Chr. 12: 10801532 - 10802627 on Build GRCh38
UniGene ID
Hs.533754
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023919.2 NP_076408.1 1 160
GenBank mRNA BC069091.1 - 1 103
BC113561.1 - 1 160 8322613

View Full Product Details