Gene Transcripts

Gene Symbol
NCKIPSD
Entrez Gene ID
51517
Gene Name
NCK interacting protein with SH3 domain
Gene Aliases
AF3P21, DIP, DIP1, ORF1, SPIN90, VIP54, WASLBP, WISH
Chromosome Location
Chr. 3: 48663814 - 48685941 on Build GRCh38
UniGene ID
Hs.655006
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016453.3 NP_057537.1 12 2045
NM_184231.2 NP_909119.1 12 2024
XM_017006594.1 XP_016862083.1 12 2039
XM_017006595.1 XP_016862084.1 12 2032
XM_017006596.1 XP_016862085.1 12 2059
XM_017006597.1 XP_016862086.1 12 1803
XM_017006598.1 XP_016862087.1 12 2029
XM_017006599.1 XP_016862088.1 12 2065
XM_017006600.1 XP_016862089.1 12 2028
GenBank mRNA AF178432.1 - 12 2013
AF303581.1 - 12 1984
AJ242655.2 - 12 2013
AK294151.1 - 4 664
AY453794.1 - 12 1992
BC016052.1 - 12 1908 4862348
BC026280.1 - 12 1975 4730420

View Full Product Details