Gene Transcripts

Gene Symbol
UBE2D4
Entrez Gene ID
51619
Gene Name
ubiquitin conjugating enzyme E2 D4 (putative)
Gene Aliases
HBUCE1
Chromosome Location
Chr. 7: 43926436 - 43956136 on Build GRCh38
UniGene ID
Hs.19196
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015983.3 NP_057067.1 3, 4 228
XM_006715734.3 XP_006715797.1 3, 4 228
XM_011515420.2 XP_011513722.1 4, 5 321
XM_011515422.2 XP_011513724.1 2, 3 165
XM_017012292.1 XP_016867781.1 4, 5 321
XM_017012293.1 XP_016867782.1 4, 5 321
XM_017012294.1 XP_016867783.1 4, 5 321
XM_017012295.1 XP_016867784.1 1, 2 75
GenBank mRNA AF125044.1 - 3, 4 267
AF125045.1 - 2, 3 114
AK001446.1 - 3, 4 216
BC004104.2 - 3, 4 220 3835609
BG499454.1 - 3, 4 243

View Full Product Details