Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018014.3 | NP_060484.2 | 2 | 429 | |
NM_022893.3 | NP_075044.2 | 2 | 429 | ||
NM_138559.1 | NP_612569.1 | 2 | 429 | ||
XM_011532909.1 | XP_011531211.1 | 2 | 429 | ||
XM_011532910.1 | XP_011531212.1 | 2 | 429 | ||
XM_011532912.1 | XP_011531214.1 | 2 | 429 | ||
XM_017004333.1 | XP_016859822.1 | 2 | 197 | ||
XM_017004334.1 | XP_016859823.1 | 2 | 429 | ||
XM_017004335.1 | XP_016859824.1 | 2 | 239 | ||
GenBank mRNA | AB058712.1 | - | 2 | 329 | |
AF080216.1 | - | 3 | 542 | ||
AJ404611.1 | - | 2 | 429 | ||
AJ404612.1 | - | 2 | 429 | ||
AJ404613.1 | - | 2 | 429 | ||
AK055251.1 | - | 2 | 429 | ||
AK300012.1 | - | 2 | 438 | ||
AY228763.1 | - | 2 | 362 | ||
AY692278.1 | - | 2 | 201 | ||
BC021098.1 | - | 2 | 310 | 5087967 |