Gene Transcripts

Gene Symbol
SH3TC1
Entrez Gene ID
54436
Gene Name
SH3 domain and tetratricopeptide repeats 1
Gene Aliases
Chromosome Location
Chr. 4: 8182072 - 8241103 on Build GRCh38
UniGene ID
Hs.479116
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318480.1 NP_001305409.1 12 2107
NM_018986.4 NP_061859.3 12 2024
NR_134639.1 10 1738
XM_006713889.2 XP_006713952.1 9 1442
XM_006713891.3 XP_006713954.1 6 1084
XM_011513488.1 XP_011511790.1 12 2001
XM_011513489.2 XP_011511791.1 10 1624
XM_017008300.1 XP_016863789.1 12 2133
XM_017008301.1 XP_016863790.1 12 1816
XM_017008302.1 XP_016863791.1 9 1526
GenBank mRNA AK000363.1 - 8 1412
AK074093.1 - 11 1863
AK074402.1 - 12 1901
AK126165.1 - 12 1980
AK222865.1 - 8 1412

View Full Product Details