Gene Transcripts

Gene Symbol
ROBO4
Entrez Gene ID
54538
Gene Name
roundabout guidance receptor 4
Gene Aliases
ECSM4, MRB
Chromosome Location
Chr. 11: 124884218 - 124897935 on Build GRCh38
UniGene ID
Hs.524121
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301088.1 NP_001288017.1 17 2889
NM_019055.5 NP_061928.4 17 3095
XM_006718861.1 XP_006718924.1 17 2914
XM_011542875.1 XP_011541177.1 11 1857
GenBank mRNA AF361473.1 - 17 3024
AK025195.1 - 5 945
AK027852.1 - 17 2993
AK074163.1 - 15 3826
AK289769.1 - 17 2994
AK297845.1 - 11 1675
AK302628.1 - 17 2759
AY358083.1 - 17 2946
BC014995.1 - 16 2707
BC025675.1 - 4 2418
BC039602.1 - 17 2983 5590503
BC064643.1 - 7 1242
BC111562.1 - 17 2989 40068748
BC111748.1 - 17 2989 40068752

View Full Product Details