Gene Transcripts

Gene Symbol
DCHS2
Entrez Gene ID
54798
Gene Name
dachsous cadherin-related 2
Gene Aliases
CDH27, CDHJ, CDHR7, PCDH23, PCDHJ
Chromosome Location
Chr. 4: 154234375 - 154491778 on Build GRCh38
UniGene ID
Hs.655664
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017639.3 NP_060109.2 18 4381
XM_011532045.1 XP_011530347.1 13 6169
XM_011532046.2 XP_011530348.1 12 3825
XM_011532049.2 XP_011530351.1 5 1675
GenBank mRNA AY354497.1 - 18 4381

View Full Product Details