Gene Transcripts

Gene Symbol
RNF43
Entrez Gene ID
54894
Gene Name
ring finger protein 43
Gene Aliases
RNF124, URCC
Chromosome Location
Chr. 17: 58352500 - 58418895 on Build GRCh38
UniGene ID
Hs.584916
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001305544.1 NP_001292473.1 8 1632
NM_001305545.1 NP_001292474.1 7 714
NM_017763.5 NP_060233.3 8 1835
XM_011524955.2 XP_011523257.1 8 1734
XM_011524956.2 XP_011523258.1 7 2511
XM_017024800.1 XP_016880289.1 9 3061
GenBank mRNA AB081837.1 - 8 1372
AK000322.1 - 7 1052
AK291027.1 - 8 1605
AK296769.1 - 6 825
AK298789.1 - 3 245
AK299024.1 - 7 714
AK309297.1 - 7 701
BC109028.2 - 7 1081 40027722
CR627423.1 - 8 1632
CR749257.1 - 7 2840

View Full Product Details