Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_017956.3 | NP_060426.2 | 1 | 1121 | |
GenBank mRNA | AK000779.1 | - | 1 | 1109 | |
AK222727.1 | - | 1 | 1109 | ||
AK225083.1 | - | 1 | 1108 | ||
BC003057.2 | - | 1 | 1066 | ||
BC011713.2 | - | 1 | 1096 | 3929724 |