Gene Transcripts

Gene Symbol
WDR70
Entrez Gene ID
55100
Gene Name
WD repeat domain 70
Gene Aliases
Chromosome Location
Chr. 5: 37379280 - 37752672 on Build GRCh38
UniGene ID
Hs.213690
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018034.2 NP_060504.1 11 1162
XM_005248318.3 XP_005248375.1 11 1180
XM_011514061.2 XP_011512363.1 5 475
XM_017009604.1 XP_016865093.1 10 996
XM_017009605.1 XP_016865094.1 10 963
GenBank mRNA AK001095.1 - 11 1104
AK291179.1 - 11 1184
AK310901.1 - 12 1285
AL512685.1 - 11 1163
BC009648.1 - 11 1162 3454326
BC025315.1 - 11 1140 5458850
BX537655.1 - 10 1119

View Full Product Details