Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_018270.5 | NP_060740.1 | 3 | 435 | |
NR_136405.1 | 3 | 435 | |||
GenBank mRNA | AB085682.1 | - | 3 | 335 | |
AK001776.1 | - | 3 | 387 | ||
BC009889.2 | - | 3 | 354 | 4026378 | |
BC014235.2 | - | 3 | 359 | 4555060 | |
BC018841.2 | - | 3 | 354 | 3050866 | |
BM920599.1 | - | 341 | |||
BU902487.1 | - | 3 | 417 |