Gene Transcripts

Gene Symbol
WDR11
Entrez Gene ID
55717
Gene Name
WD repeat domain 11
Gene Aliases
BRWD2, DR11, HH14, SRI1, WDR15
Chromosome Location
Chr. 10: 120851175 - 120909526 on Build GRCh38
UniGene ID
Hs.144447
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018117.11 NP_060587.8 24 3264
XM_005269962.2 XP_005270019.1 24 2981
XM_005269963.2 XP_005270020.1 24 3113
XM_005269964.2 XP_005270021.1 17 2019
XM_017016397.1 XP_016871886.1 23 2952
XM_017016398.1 XP_016871887.1 23 2882
XM_017016399.1 XP_016871888.1 23 2826
GenBank mRNA AB037772.2 - 24 3077
AF320223.1 - 24 3051
AK000579.1 - 10 1066
AK001368.1 - 19 2167
AK025050.1 - 7 775
AK124522.1 - 11 3451
AK299524.1 - 17 1884
AK301761.1 - 4 464
AK315145.1 - 24 3042
AL137699.1 - 22 2737
BC040469.1 - 24 3081 4730308
BC071564.1 - 24 3047 30346203

View Full Product Details