Gene Transcripts
- Gene Symbol
- EXOC2
- Entrez Gene ID
- 55770
- Gene Name
- exocyst complex component 2
- Gene Aliases
- SEC5, SEC5L1, Sec5p
- Chromosome Location
- Chr. 6: 485138 - 693141 on Build GRCh38
- UniGene ID
- Hs.484412
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_018303.5 |
NP_060773.3
|
15, 16 |
1841 |
|
NR_073064.1 |
|
17, 18 |
2003 |
|
XM_017011018.1 |
XP_016866507.1
|
16, 17 |
1887 |
|
XM_017011019.1 |
XP_016866508.1
|
16, 17 |
2049 |
|
XM_017011020.1 |
XP_016866509.1
|
16, 17 |
1909 |
|
XM_017011021.1 |
XP_016866510.1
|
15, 16 |
2145 |
|
XM_017011022.1 |
XP_016866511.1
|
15, 16 |
1841 |
|
XM_017011023.1 |
XP_016866512.1
|
15, 16 |
1841 |
|
XM_017011024.1 |
XP_016866513.1
|
16, 17 |
1887 |
|
XM_017011025.1 |
XP_016866514.1
|
15, 16 |
1841 |
|
XM_017011026.1 |
XP_016866515.1
|
15, 16 |
1841 |
|
GenBank mRNA |
AJ414403.1 |
- |
15, 16 |
1765 |
|
AK001888.1 |
- |
11, 12 |
1256 |
|
AK056564.1 |
- |
15, 16 |
1809 |
|
AK302941.1 |
- |
5, 6 |
582 |
|
AK314628.1 |
- |
15, 16 |
1841 |
|
AL833209.1 |
- |
13, 14 |
1397 |
|
AL833213.1 |
- |
15, 16 |
1787 |
|
BC016918.1 |
- |
12, 13 |
1321 |
|
BC080542.1 |
- |
17, 18 |
1973 |
5479496 |
View Full Product Details