Gene Transcripts
- Gene Symbol
- TEX2
- Entrez Gene ID
- 55852
- Gene Name
- testis expressed 2
- Gene Aliases
- HT008, TMEM96
- Chromosome Location
- Chr. 17: 64147433 - 64263323 on Build GRCh38
- UniGene ID
- Hs.175414
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001288732.1 |
NP_001275661.1
|
10, 11 |
3314 |
|
NM_001288733.1 |
NP_001275662.1
|
10, 11 |
3306 |
|
NM_018469.4 |
NP_060939.3
|
10, 11 |
3335 |
|
XM_011524998.1 |
XP_011523300.1
|
10, 11 |
3324 |
|
XM_011524999.1 |
XP_011523301.1
|
10, 11 |
3248 |
|
XM_011525000.1 |
XP_011523302.1
|
10, 11 |
3299 |
|
XM_017024846.1 |
XP_016880335.1
|
10, 11 |
3278 |
|
GenBank mRNA |
AB051525.2 |
- |
9, 10 |
3133 |
|
AF163260.1 |
- |
8, 9 |
1351 |
|
AF220182.1 |
- |
6, 7 |
869 |
|
AK000369.1 |
- |
5, 6 |
640 |
|
AK000758.1 |
- |
9, 10 |
1550 |
|
AK091156.1 |
- |
9, 10 |
2303 |
|
AK314316.1 |
- |
5, 6 |
585 |
|
AL832371.1 |
- |
10, 11 |
3214 |
|
AL834251.2 |
- |
10, 11 |
3225 |
|
BC008896.2 |
- |
9, 10 |
2490 |
|
BC033661.1 |
- |
7, 8 |
1017 |
|
BC036672.1 |
- |
10, 11 |
3285 |
|
BC040521.1 |
- |
10, 11 |
3239 |
4837394 |
BC040522.1 |
- |
10, 11 |
3287 |
|
BC108279.1 |
- |
8, 9 |
1372 |
|
BX648260.1 |
- |
2, 3 |
1596 |
|
CR627433.1 |
- |
10, 11 |
3307 |
|
View Full Product Details