Gene Transcripts

Gene Symbol
KIZ
Entrez Gene ID
55857
Gene Name
kizuna centrosomal protein
Gene Aliases
C20orf19, HT013, Kizuna, NCRNA00153, PLK1S1, RP69
Chromosome Location
Chr. 20: 21125980 - 21246628 on Build GRCh38
UniGene ID
Hs.187635
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163022.1 NP_001156494.1 4 895
NM_001163023.1 NP_001156495.1 3 805
NM_001276389.1 NP_001263318.1 3 832
NM_018474.4 NP_060944.3 5 1058
XM_011529296.2 XP_011527598.1 5 1061
XM_011529297.2 XP_011527599.1 5 1050
XM_011529298.2 XP_011527600.1 5 1043
XM_011529299.2 XP_011527601.1 4 890
XM_017027951.1 XP_016883440.1 6 1004
XM_017027952.1 XP_016883441.1 4 890
XM_017027953.1 XP_016883442.1 6 1076
XM_017027954.1 XP_016883443.1 4 890
GenBank mRNA AF220187.1 - 1 90
AF451990.1 - 5 1035
AK025476.1 - 5 989
AK293166.1 - 3 736
AK293567.1 - 3 776
AK295355.1 - 4 794
AK296399.1 - 1 324
AL110196.1 - 1 337
BC039296.1 - 5 1019
BC045826.1 - 4 782 5262165
BC065550.1 - 3 639
BC090879.2 - 1 463
BC105093.1 - 5 961 8144096
BC113370.1 - 5 961 8322422

View Full Product Details