Gene Transcripts

Gene Symbol
ZMAT5
Entrez Gene ID
55954
Gene Name
zinc finger matrin-type 5
Gene Aliases
SNRNP20, U11/U12-20K, ZC3H19
Chromosome Location
Chr. 22: 29730956 - 29767011 on Build GRCh38
UniGene ID
Hs.713647
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003692.1 NP_001003692.1 2 254
NM_001318129.1 NP_001305058.1 2 208
NM_019103.2 NP_061976.1 3 369
GenBank mRNA AA312132.1 - 2 254
AK292671.1 - 2 235
AL160311.1 - 2 253
BC009717.1 - 2 219 3890730
BG285784.1 - 3 333
BQ931527.1 - 2 182
BU598307.1 - 2 256
CR456353.1 - 1 141
CR457393.1 - 1 118
DT217179.1 - 3 385

View Full Product Details