Gene Transcripts
- Gene Symbol
- XAB2
- Entrez Gene ID
- 56949
- Gene Name
- XPA binding protein 2
- Gene Aliases
- HCNP, HCRN, NTC90, SYF1
- Chromosome Location
- Chr. 19: 7619525 - 7629553 on Build GRCh38
- UniGene ID
- Hs.9822
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_020196.2 |
NP_064581.2
|
8, 9 |
1149 |
|
GenBank mRNA |
AB026111.1 |
- |
8, 9 |
1138 |
|
AB033003.1 |
- |
6, 7 |
825 |
|
AF226051.1 |
- |
8, 9 |
1148 |
|
AF258567.1 |
- |
8, 9 |
1144 |
|
AF272147.1 |
- |
8, 9 |
1159 |
|
AK025858.1 |
- |
3, 4 |
432 |
|
AK074035.1 |
- |
4, 5 |
1720 |
|
BC007208.1 |
- |
8, 9 |
1131 |
2823235 |
BC008778.1 |
- |
3, 4 |
464 |
|
CR749864.1 |
- |
5, 6 |
678 |
|
View Full Product Details