Gene Transcripts

Gene Symbol
EPB41L5
Entrez Gene ID
57669
Gene Name
erythrocyte membrane protein band 4.1 like 5
Gene Aliases
BE37, YMO1, YRT
Chromosome Location
Chr. 2: 120013028 - 120179121 on Build GRCh38
UniGene ID
Hs.369232
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001184937.1 NP_001171866.1 17 1584
NM_020909.3 NP_065960.2 17 1584
XM_006712651.1 XP_006712714.1 17 1564
XM_006712652.1 XP_006712715.1 17 1879
XM_006712653.1 XP_006712716.1 17 1584
XM_011511529.2 XP_011509831.1 17 1564
XM_017004567.1 XP_016860056.1 17 1420
GenBank mRNA BC054508.1 - 17 1519

View Full Product Details