Gene Transcripts

Gene Symbol
SLC7A14
Entrez Gene ID
57709
Gene Name
solute carrier family 7 member 14
Gene Aliases
PPP1R142
Chromosome Location
Chr. 3: 170459554 - 170586074 on Build GRCh38
UniGene ID
Hs.596660
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020949.2 NP_066000.2 2 449
GenBank mRNA AK122655.1 - 2 444
BC022968.1 - 1 65
DA475001.1 - 2 449

View Full Product Details