Gene Transcripts

Gene Symbol
SUDS3
Entrez Gene ID
64426
Gene Name
SDS3 homolog, SIN3A corepressor complex component
Gene Aliases
SAP45, SDS3
Chromosome Location
Chr. 12: 118373189 - 118418035 on Build GRCh38
UniGene ID
Hs.416630
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022491.2 NP_071936.2 3, 4 405
XM_011538671.2 XP_011536973.1 4, 5 1482
XM_017019817.1 XP_016875306.1 2, 3 289
XM_017019818.1 XP_016875307.1 5, 6 637
XM_017019819.1 XP_016875308.1 4, 5 499
XM_017019820.1 XP_016875309.1 3, 4 229
GenBank mRNA AK024460.1 - 3, 4 388
BC098404.1 - 3, 4 428 30332266
BX647687.1 - 4, 5 363

View Full Product Details