Gene Transcripts

Gene Symbol
C7orf49
Entrez Gene ID
78996
Gene Name
chromosome 7 open reading frame 49
Gene Aliases
MRI
Chromosome Location
Chr. 7: 135092303 - 135170826 on Build GRCh38
UniGene ID
Hs.643113
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001243749.1 NP_001230678.1 3, 4 821
NM_001243751.1 NP_001230680.1 3, 4 568
NM_001243752.1 NP_001230681.1 3, 4 642
NM_001243753.1 NP_001230682.1 3, 4 446
NM_001243754.1 NP_001230683.1 3, 4 348
NM_001243755.1 NP_001230684.1 3, 4 264
NM_024033.3 NP_076938.2 3, 4 531
XM_017012589.1 XP_016868078.1 3, 4 654
XM_017012590.1 XP_016868079.1 3, 4 580
XM_017012591.1 XP_016868080.1 3, 4 434
XM_017012593.1 XP_016868082.1 3, 4 484
XM_017012594.1 XP_016868083.1 3, 4 691
GenBank mRNA AK026103.1 - 3, 4 657
AK130795.1 - 1, 2 357
AL708733.1 - 3, 4 264
BC000168.2 - 3, 4 400 2900206
BC067350.1 - 3, 4 437 6568468
BG770658.1 - 3, 4 431
BM727569.1 - 2, 3 362
BP213903.1 - 3, 4 429
BQ423284.1 - 281
DA213716.1 - 3, 4 525
DB129229.1 - 3, 4 522

View Full Product Details