Gene Transcripts
- Gene Symbol
- SECISBP2
- Entrez Gene ID
- 79048
- Gene Name
- SECIS binding protein 2
- Gene Aliases
- SBP2
- Chromosome Location
- Chr. 9: 89318473 - 89367117 on Build GRCh38
- UniGene ID
- Hs.59804
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001282688.1 |
NP_001269617.1
|
6, 7 |
989 |
|
NM_001282689.1 |
NP_001269618.1
|
6, 7 |
871 |
|
NM_001282690.1 |
NP_001269619.1
|
6, 7 |
1069 |
|
NM_024077.4 |
NP_076982.3
|
6, 7 |
992 |
|
XM_006717282.2 |
XP_006717345.1
|
6, 7 |
925 |
|
XM_011519000.2 |
XP_011517302.1
|
6, 7 |
1046 |
|
XM_011519001.1 |
XP_011517303.1
|
6, 7 |
1046 |
|
XM_017015122.1 |
XP_016870611.1
|
6, 7 |
959 |
|
GenBank mRNA |
AF380995.1 |
- |
6, 7 |
945 |
|
AK090608.1 |
- |
6, 7 |
959 |
|
AK290182.1 |
- |
6, 7 |
838 |
|
AK299161.1 |
- |
6, 7 |
1080 |
|
AK302852.1 |
- |
6, 7 |
987 |
|
AL136881.1 |
- |
6, 7 |
825 |
|
BC036109.1 |
- |
6, 7 |
954 |
5296292 |
BC068466.1 |
- |
6, 7 |
927 |
|
View Full Product Details