Gene Transcripts

Gene Symbol
OR52N1
Entrez Gene ID
79473
Gene Name
olfactory receptor family 52 subfamily N member 1
Gene Aliases
OR11-61
Chromosome Location
Chr. 11: 5787854 - 5788816 on Build GRCh38
UniGene ID
Hs.690209
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001913.1 NP_001001913.1 1 281

View Full Product Details